Orders LinkContactUsLinkComponent
Shelving systems, solid, Super Erecta®
Super Erecta® solid flat shelf, (width×length): 457×1219 mm, 1848FS, Acciaio inossidabile 304, 9,1 kg
  139-0320
Shelving systems, solid, Super Erecta®
  139-0320
Shelving systems, solid, Super Erecta®
Shelving systems, solid, Super Erecta®
Super Erecta® solid flat shelf, (width×length): 457×1219 mm, 1848FS, Acciaio inossidabile 304, 9,1 kg
SFr. 701.00
1 * 1 ST
Perfect for applications involving spillage. Provides barrier between floor and bottom shelf contents to deter contamination.
  • Shelf design features a 3,2 mm raised 'ship’s edge' on all four sides to contain spills
  • Louvered/embossed shelf allows air circulation
  • Available in several styles and materials for a variety of applications
  • Shelves are constructed with 18-gauge stainless steel (type 304) or galvanized materials
Standard stainless steel solid shelves (Type 304): with epoxy-coated cast corners address the majority of applications for solid, corrosion resistant shelves or work surfaces. Available in flat solid and louvered/embossed styles.
Galvanized shelves: with uncoated cast corners are ideal for applications requiring a solid shelving or work surface and minimum resistance to corrosion. Available in flat solid and louvered/embossed styles.
Premium Autoclave/Cart Washable Stainless Steel solid shelves (Type 304): feature an all-stainless construction to withstand high temperature, corrosive applications. Available in flat solid style.
 : Each individual shelf and post is sold separately. Accessories are also available.
  • Descrizione:
    Super Erecta® solid flat shelf, (width×length): 457×1219 mm, 1848FS
  • Materiale:
    Acciaio inossidabile 304
  • Seq:
    14
  • Peso:
    9,1 kg
Accessories
  •  
    Descrizione
  • METO9990P
    Label holders, for 75×32 mm labels
    1 * 1 ST - SFr. 8.90
  •  
    Descrizione
    Per
  • 139-0268
    Montante mobile, SiteSelect™, cromato, altezza 1590 mm
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    1 * 1 ST - SFr. 53.80
  •  
    Descrizione
    Per
  • 139-0270
    Montante mobile, SiteSelect™, cromato, altezza 2454 mm
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    1 * 1 ST - SFr. 74.80
  •  
    Descrizione
    Per
  • 139-0277
    Montante mobile, SiteSelect™, in acciaio inox, altezza 2454 mm
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale Super Erecta Pro™
    1 * 1 ST - SFr. 202.00
  •  
    Descrizione
  • METO9990SL
    Slanted label holders, for 75×32 mm labels
    1 * 1 ST - SFr. 8.90
  •  
    Descrizione
    Per
  • 139-0275
    Montante mobile, SiteSelect™, in acciaio inox, altezza 1590 mm
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale Super Erecta Pro™
    1 * 1 ST - SFr. 149.00
  •  
    Descrizione
    Per
  • 139-0255
    Montante fisso, SiteSelect™, cromato, altezza 2200 mm
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    1 * 1 ST - SFr. 74.80
  •  
    Descrizione
    Per
  • 139-0251
    Montante fisso, SiteSelect™, cromato, altezza 875 mm
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    1 * 1 ST - SFr. 45.70
  •  
    Descrizione
  • METO2424BWI
    Decorator shelf inlays, black/white, 610×610 mm
    1 * 1 ST - SFr. 76.60
  •  
    Descrizione
  • METO9985
    Plastic split sleeves, black
    1 * 4 ST - SFr. 16.10
  •  
    Descrizione
  • METO9990CL5
    Clear label holders,1520 mm, for 1395×32 mm labels
    1 * 1 ST - SFr. 45.10
  •  
    Descrizione
    Per
  • 139-0269
    Montante mobile, SiteSelect™, cromato, altezza 1880 mm
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    1 * 1 ST - SFr. 58.70
  •  
    Descrizione
  • METO9986S
    Aluminum split sleeves with stainless rings
    1 * 1 Un. - SFr. 129.00
  •  
    Descrizione
    Per
  • 139-0276
    Montante mobile, SiteSelect™, in acciaio inox, altezza 1880 mm
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale Super Erecta Pro™
    1 * 1 ST - SFr. 164.00
  •  
    Descrizione
    Per
  • 139-0260
    Montante fisso, SiteSelect™, in acciaio inox, altezza 1590 mm
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale fisso, scaffale Super Erecta Pro™, scaffale Super Erecta Pro™
    1 * 1 ST - SFr. 149.00
  •  
    Descrizione
    Per
  • METO63PK3
    Montante fisso, SiteSelect™, metroseal 3, altezza 1590 mm
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta Pro™
    1 * 1 ST - SFr. 63.00
  •  
    Descrizione
  • METOEP36C
    Enclosure panel, chrome, 313×1518 mm, for nominal post height 1600 mm
    1 * 1 ST - SFr. 143.00
  •  
    Descrizione
    Per
  • 139-0253
    Montante fisso, SiteSelect™, cromato, altezza 1590 mm
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    1 * 1 ST - SFr. 53.80
  •  
    Descrizione
    Per
  • METO74PK3
    Montante fisso, SiteSelect™, metroseal 3, altezza 1895 mm
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta Pro™
    1 * 1 ST - SFr. 68.60
  •  
    Descrizione
  • METO2WS21C
    Post mounting shelf supports, width 530 mm, double, chrome plated
    1 * 1 ST - SFr. 128.00
  •  
    Descrizione
    Per
  • 139-0254
    Montante fisso, SiteSelect™, cromato, altezza 1895 mm
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    1 * 1 ST - SFr. 58.70
  •  
    Descrizione
  • METO2WS14C
    Post mounting shelf supports, width 355 mm, double, chrome plated
    1 * 1 ST - SFr. 122.00
  •  
    Descrizione
  • METO1WS21C
    Post mounting shelf supports, width 530 mm, single, chrome plated
    1 * 1 ST - SFr. 85.90
  •  
    Descrizione
  • METO1WS14C
    Post mounting shelf supports, width 355 mm, single, chrome plated
    1 * 1 ST - SFr. 82.80
  •  
    Descrizione
  • METOH212C
    Storage baskets, chrome plated, 190×440×225 mm, 5 kg
    1 * 1 ST - SFr. 114.00
  •  
    Descrizione
  • METOH209C
    Storage baskets, chrome plated, 127×345×180 mm, 4 kg
    1 * 1 ST - SFr. 81.60
  •  
    Descrizione
  • METOEP57C
    Enclosure panel, chrome, 464×1791 mm, for nominal post height 1800 mm
    1 * 1 ST - SFr. 179.00
  •  
    Descrizione
  • METO9084Z
    Additional tabs for rod, bag of 12
    1 * 1 Un. - SFr. 20.40
  •  
    Descrizione
    Materiale
  • METO1436CI-4
    Clear shelf inlays for Super Erecta® shelving system, 914×355 mm
    Plastica
    1 * 1 Un. - SFr. 147.00
  •  
    Descrizione
  • METOH118C
    Hanger rail fits on posts along the width or length of the unit 457 mm, fits to size shelf width: 914 mm. Optional hooks can hang from rail at any point.
    1 * 1 ST - SFr. 43.30
  •  
    Descrizione
  • METO9986Z
    Aluminum split sleeves with zinc rings perfect for high temperature or conductive applications ; come with zinc retainer rings to secure the shelf in place (one pack required/shelf)
    1 * 1 Un. - SFr. 21.70
  •  
    Descrizione
  • METOHHK6C
    Extension display hanger (152 mm): snaps on Super Erecta® shelf hanger rails and shelf frames, accepts standard carded merchandise
    1 * 1 ST - SFr. 22.90
  •  
    Descrizione
  • METO9985W
    Split sleeves, plastic, white
    1 * 1 Un. - SFr. 16.10
  •  
    Descrizione
  • METO9990CL
    Slanted label holders, for 75×32 mm labels
    1 * 1 ST - SFr. 8.90
  •  
    Descrizione
  • METO9990CL3
    Clear label holders, plastic, fits 914 mm shelfs, 725×32 mm
    1 * 1 ST - SFr. 26.60
  •  
    Descrizione
    Materiale
  • METO2136CI-4
    Clear shelf inlays for Super Erecta® shelving system, 914×530 mm
    Plastica
    1 * 1 Un. - SFr. 159.00
  •  
    Descrizione
    Per
  • METO63UPS-SW
    Swaged post, stainless steel with aluminium cap, 1600 mm dimension
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta Pro™ shelving
    1 * 1 ST - SFr. 164.00
  •  
    Descrizione
    Per
  • METO96P
    Stationary post, SiteSelect™, chrome plated, 2454 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 ST - SFr. 85.30
  •  
    Descrizione
    Per
  • 139-0272
    Mobile post, SiteSelect™, stainless steel, 699 mm height
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 ST - SFr. 106.00
  •  
    Descrizione
    Per
  • 139-0258
    Stationary post, SiteSelect™, stainless steel, 875 mm height
    Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 ST - SFr. 120.00
  •  
    Descrizione
    Per
  • 139-0257
    Stationary post, SiteSelect™, stainless steel, 875 mm height
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 ST - SFr. 106.00
  •  
    Descrizione
    Per
  • 139-0267
    Mobile post, SiteSelect™, chrome plated, 1385 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 ST - SFr. 51.30
  •  
    Descrizione
    Per
  • 139-0273
    Mobile post, SiteSelect™, stainless steel, 857 mm height
    Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 ST - SFr. 120.00
  •  
    Descrizione
    Per
  • 139-0252
    Stationary post, SiteSelect™, chrome plated, 1385 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
    1 * 1 ST - SFr. 51.30
  •  
    Descrizione
    Per
  • 139-0262
    Stationary post, SiteSelect™, stainless steel, 2200 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 ST - SFr. 193.00
  •  
    Descrizione
    Per
  • 139-0263
    Mobile post, SiteSelect™, chrome plated, 191 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 ST - SFr. 31.50
  •  
    Descrizione
    Per
  • 139-0248
    Stationary post, SiteSelect™, chrome plated, 191 mm height
    Drop Mat™ Super Erecta® display shelving, Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, APPEAL™ display shelving
    1 * 1 ST - SFr. 31.50
  •  
    Descrizione
    Per
  • 139-0274
    Mobile post, SiteSelect™, stainless steel, 1385 mm height
    Super Adjustable Super Erecta® shelving, Super Erecta® shelving, Super Erecta® Solid shelving, Super Erecta Pro™ shelving
    1 * 1 ST - SFr. 131.00
  •  
    Descrizione
    Per
  • 139-0271
    Mobile post, SiteSelect™, stainless steel, 370 mm height
    Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving
    1 * 1 ST - SFr. 79.70
  •  
    Descrizione
    Per
  • 139-0264
    Mobile post, SiteSelect™, chrome plated, 370 mm height
    Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving
    1 * 1 ST - SFr. 35.20
  •  
    Descrizione
    Per
  • 139-0256
    Stationary post, SiteSelect™, stainless steel, 370 mm height
    Super Erecta Pro™, Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® solid shelving
    1 * 1 ST - SFr. 79.70
  •  
    Descrizione
    Per
  • 139-0261
    Montante fisso, SiteSelect™, in acciaio inox, altezza 1895 mm
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale fisso, scaffale Super Erecta Pro™
    1 * 1 ST - SFr. 164.00
  •  
    Descrizione
  • METO9994Z
    Post clamps, joins units together for maximum strength, zinc-plated
    1 * 1 ST - SFr. 15.50
  •  
    Descrizione
    Per
  • METO74UPK3
    Montante mobile, SiteSelect™, metroseal 3, altezza 1880 mm
    Scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta Pro™
    1 * 1 ST - SFr. 68.60
  •  
    Descrizione
    Per
  • 139-0265
    Montante mobile, SiteSelect™, cromato, altezza 699 mm
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    1 * 1 ST - SFr. 42.70
  •  
    Descrizione
    Per
  • 139-0259
    Stationary post, SiteSelect™, stainless steel, 1385 mm height
    Super Erecta Pro™ and Super Erecta® shelving, Super Adjustable Super Erecta® shelving and Super Erecta® Solid shelving
    1 * 1 ST - SFr. 131.00
  •  
    Descrizione
    Per
  • 139-0250
    Montante fisso, SiteSelect™, cromato, altezza 699 mm
    Scaffale aperto Drop Mat™ Super Erecta®, scaffale Super Adjustable Super Erecta®, scaffale Super Erecta®, scaffale Super Erecta® Solid, scaffale aperto APPEAL™
    1 * 1 ST - SFr. 42.70
  •  
    Descrizione
  • METO9986HZ
    Heavy duty aluminum split sleeves with zinc rings
    1 * 1 ST - SFr. 56.90