Labeling tapes, warning tapes
Warning tape, self-adhesive PVC, 0,138 mm thick, nero/giallo, 50,0 mm
Codice catalogo BRDY255312
Nome del fornitore: Brady
Vai a
Product Details & Documents
Self-adhesive warning film for marking pillars, pylons, the edges of walls, gate arches and lifts.
- Extremely durable
- Waterproof, oil, alkali and acid resistant
- Different colours available
Length: 33,0 m.
Specifiche
- Larghezza:50,0 mm
- Colore:nero/giallo
- Descrizione:Warning tape, self-adhesive PVC, 0,138 mm thick
- Seq:02