Labels, hazard warning
Hazard labels: danger asbestos, —, Poliestere, nero on giallo, 26×52 mm, 250 labels/roll
Codice catalogo BRDY223335
Nome del fornitore: Brady
Vai a
Product Details & Documents
These self adhesive signs are designed to warn people to be careful and to take precautions of potential hazards.
Available in different sizes.
Certifications: Conforms to Health and Safety (Safety Signs and Signals) Regulations 1996.
Documents
Specifiche
- Colore:nero on giallo
- Descrizione:Hazard labels: danger asbestos
- Dimensioni:26×52 mm
- Materiale:Poliestere
- Seq:07
- Modello:—
- Confezione:250 labels/roll
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.