Sample containers, with screw cap, universal, WHEATON®
30 ml, Universal container, printed label, sterile, traslucido, PP/PE
Codice catalogo 216-0987
Nome del fornitore: DWK Life Sciences
Sample containers, with screw cap, universal, WHEATON®
Codice catalogo 216-0987
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
PP translucent or PS clear, with PE screw cap.
- Manufactured from FDA approved virgin polystyrene or polypropylene
- PS offers good thermal and chemical resistance
- Supplied sterile - for in vitro use only
- All materials are non cytotoxic
Straight walled containers ideal for sample collection, storage and transport. Supplied with leakproof, flow seal closure. Available pre-filled with boric acid for urine samples.
Certifications: CE marked in accordance with European Directive 98/79/EC
All containers are leak tested to British Standard BS5213:1975(1989)
Confezione: Packed in inner bags of 50.
Attenzione: *I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904
Specifiche
- Tipo:Universal container, printed label, sterile
- Capacità:30 ml
- Colore:traslucido
- Materiale:PP/PE