ALUMINUM HEATING BLOCK FOR 96 WELL INSERTS, 1.0ML & 1.2ML HEATING BLOCK FOR 1.0ML INS 1 * 1 items
Catalog # MLAS07-HTGB-1000
Supplier: MICROLITER ANALYTICAL SUPPLIES
ALUMINUM HEATING BLOCK FOR 96 WELL INSERTS, 1.0ML & 1.2ML HEATING BLOCK FOR 1.0ML INS 1 * 1 items
Catalog # MLAS07-HTGB-1000
Supplier: MICROLITER ANALYTICAL SUPPLIES
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Metanolo ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., grado gradiente per HPLC
Supplier: VWR Chemicals
Acqua, HiPerSolv CHROMANORM® per HPLC
Supplier: VWR Chemicals
Guanti in nitrile, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Pipette sierologiche, Standard Line
Supplier: VWR Collection
VWR®, Guanti in Nitrile
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® per LC-MS, adatto per UPLC/UHPLC
Supplier: VWR Chemicals
VWR®, Salviette per il viso
Supplier: VWR Collection
Strisce reattive pH, MQuant®, Supelco®
Supplier: Merck
VWR® NITRILE LIGHT, Guanti in nitrile
Supplier: VWR Collection
VWR®, pipette di trasferimento, alte prestazioni
Supplier: VWR Collection