Sample containers, straight, with screw cap, WHEATON®
250 ml, Plain label, sterile, trasparente, PS/metal
Anbieter: DWK Life Sciences
Sample containers, straight, with screw cap, WHEATON®
Katalognr. 216-1020
Anbieter: DWK Life Sciences
Direkt zu
Product Details & Documents
Polystyrene clear, with metal screw cap.
- Manufactured from FDA approved virgin polystyrene
- Supplied sterile and for in vitro use only
- CE marked in accordance with European Directive 98/79/EC
- All containers are leak tested to British Standard BS5213:1975(1989)
- All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Achtung! Bitte beachten: *I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904
Dokumente
Zertifikate
Suche nach ChargennummerCoC_LSUK360200_8321118 CoC_LSUK360206_0680228 CoC_LSUK360206_1580328 coc_LSUK360207_1050316 CoC_LSUK360207_3530708
Mehr
LSUK360251_TDS_EN
Technisches Datenblatt
Technische Daten
- Tipo:Plain label, sterile
- Capacità:250 ml
- Colore:trasparente
- Materiale:PS/metal