Sample containers, straight, with screw cap, WHEATON®
250 ml, No label, sterile, trasparente, PS/metal
Nome del fornitore: DWK Life Sciences
Sample containers, straight, with screw cap, WHEATON®
Codice catalogo 216-1019
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
Polystyrene clear, with metal screw cap.
- Manufactured from FDA approved virgin polystyrene
- Supplied sterile and for in vitro use only
- CE marked in accordance with European Directive 98/79/EC
- All containers are leak tested to British Standard BS5213:1975(1989)
- All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport. Polystyrene offers good thermal and chemical resistance. Supplied with flow seal metal cap.
Attenzione: *I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904
Documents
Specifiche
- Tipo:No label, sterile
- Capacità:250 ml
- Colore:trasparente
- Materiale:PS/metal