Rain jacket, Sheffer 698Z
Verde oliva, M
Codice catalogo SIOE698Z/A41/M
Nome del fornitore: SIOEN
Vai a
Product Details & Documents
Highly breathable rain jacket made of Siopor regular fabric consisting of 100% polyester (plain weave) with water-repellent PU coating. With fixed mesh lining in body and polyester lining in sleeves.
- Straight collar with foldaway hood, zip closure under flap with press studs
- Two patched pockets
- Round inset sleeves, narrowing by press studs
- Underarm ventilation
- Zipper in back lining for placing logos
Back length: 85 cm
Weight: 155 g/m²
Certifications: EN 343 (3-3)
Specifiche
- Taglia dell'abbigliamento:M
- Colore:Verde oliva
- Seq:08
- Tessuto:Polyester/PU
- Cat. No.:SIOE698Z/A41/M
- Genere:Unisex
- Sleeve or leg length:Long
- Risk area:Rain (waterproof)
- Peso:155 g/m²
- Modello:698Z Sheffer
- Applicazioni per abiti:Outdoor
- Tipo di chiusura:Con chiusura a zip