Heparin Sodium, Reference Standards
Heparin Sodium with Oversulfated Chondroitin Sulfate, USP reference standard, Ampule, 8,8 mg
Codice catalogo 1304050.
Nome del fornitore: USP
Vai a
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifiche
Specifiche
- Tipo di conf.:Ampule
- Conf:8,8 mg
- Standard:USP reference standard
- Descrizione:Heparin Sodium with Oversulfated Chondroitin Sulfate
- Cat. No.:1304050.
- Descrizione:Heparin Sodium
- CAS No.:9041-08-1