Heparin Sodium, Reference Standards
Heparin Sodium System Suitability, USP reference standard, Vial, 50 mg
Katalognr. 1304049.
Anbieter: USP
Direkt zu
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Technische Daten
Technische Daten
- Tipo di conf.:Vial
- Conf:50 mg
- Standard:USP reference standard
- Descrizione:Heparin Sodium System Suitability
- Cat. No.:1304049.
- Formula:C₉H₂₀N₃NaO₃
- Descrizione:Heparin Sodium
- CAS No.:9041-08-1
Technische Daten
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..



