Estradiol Related Compounds, Reference Standards
Estradiol Related Compound C, USP reference standard, Vial, 15 mg
Codice catalogo 1250041.
Nome del fornitore: USP
Estradiol Related Compounds, Reference Standards
Codice catalogo 1250041.
Nome del fornitore: USP
Numero CAS:
Vai a
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifiche
Specifiche
- Tipo di conf.:Vial
- Conf:15 mg
- Standard:USP reference standard
- Descrizione:Estradiol Related Compound C
- Cat. No.:1250041.
- Formula:C₁₈H₂₂O₃
- Descrizione:Estradiol Related Compound C
- Synonym(s):1,3,5(10)-estratrien-3,17β-diol-6-one
- CAS No.:571-92-6



