Cyclophosphamide Related Compounds, Reference Standard
Cyclophosphamide Related Compound C, USP reference standard, Vial, 25 mg
Catalog # 1157035.
Supplier: USP
Jump to:
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications
Specifications
- Tipo di conf.:Vial
- Conf:25 mg
- Standard:USP reference standard
- Descrizione:Cyclophosphamide Related Compound C
- Cat. No.:1157035.
- Formula:C₃H₁₀NO₄P
- Descrizione:Cyclophosphamide Related Compound C
- Synonym(s):3-aminopropyl dihydrogen phosphate
- CAS No.:1071-28-9



