Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Amoxicillin Related Compounds, Reference Standard

Amoxicillin Related Compounds, Reference Standard

Amoxicillin Related Compound G, USP reference standard, Vial, 15 mg
Codice catalogo 1031570.
Nome del fornitore:   USP
Amoxicillin Related Compounds, Reference Standard
Codice catalogo 1031570.
Nome del fornitore:  USP
Numero CAS:  
Amoxicillin Related Compounds, Reference Standard

Amoxicillin Related Compound G, USP reference standard, Vial, 15 mg

Numero CAS:  
Price
SFr. 1’480.00
Log in per disponibilità
1 * 15 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifiche

Specifiche

  • Tipo di conf.:
    Vial
  • Conf:
    15 mg
  • Standard:
    USP reference standard
  • Descrizione:
    Amoxicillin Related Compound G
  • Cat. No.:
    1031570.
  • Formula:
    C₂₄H₂₆N₄O₇S
  • Descrizione:
    Amoxicillin Related Compound G
  • Synonym(s):
    (2S,5R,6R)-6-{(R)-2-[(R)-2-Amino-2-(4-hydroxyphenyl)acetamido]-2-(4-hydroxyphenyl)acetamido}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
  • CAS No.:
    188112-75-6

Specifiche

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..