Amoxicillin Related Compounds, Reference Standard
Amoxicillin Related Compound D, USP reference standard, Vial, 50 mg
: USP
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
- Tipo di conf.:Vial
- Conf:50 mg
- Standard:USP reference standard
- Descrizione:Amoxicillin Related Compound D
- Cat. No.:1031547.
- Formula:C₁₆H₂₀NaN₃O₆S
- Descrizione:Amoxicillin Related Compound D
- Synonym(s):(4S)-2-{[(R)-2-amino-2-(4-hydroxyphenyl)acetamido](carboxy)methyl}-5,5-dimethylthiazolidine-4-carboxylic acid, monosodium salt
- CAS No.:1642559-71-4



