Amoxicillin Related Compounds, Reference Standard
Amoxicillin Related Compound B, USP reference standard, Vial, 15 mg
Codice catalogo 1031525.
Nome del fornitore: USP
Amoxicillin Related Compounds, Reference Standard
Codice catalogo 1031525.
Nome del fornitore: USP
Numero CAS:
Vai a
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Documents
Certificates
Cerca per numero di lottoSpecifiche
Specifiche
- Tipo di conf.:Vial
- Conf:15 mg
- Standard:USP reference standard
- Descrizione:Amoxicillin Related Compound B
- Cat. No.:1031525.
- Formula:C₁₆H₁₉N₃O₅S
- Descrizione:Amoxicillin Related Compound B
- Synonym(s):L-amoxicillin
- CAS No.:26889-93-0
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



