Acebutolol, Reference Standards
Acebutolol Related Compound A, USP reference standard, 20 mg
Catalog # 1000612.
Supplier: USP
Jump to:
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications
Specifications
- Conf:20 mg
- Standard:USP reference standard
- Descrizione:Acebutolol Related Compound A
- Cat. No.:1000612.
- Descrizione:Acebutolol Related Compound A
- Synonym(s):N-(3-Acetyl-4-hydroxyphenyl)butyramide
- CAS No.:40188-45-2
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



