Robotic tips for Eppendorf epMotion® automated pipetting system
300 µl, +, -, 10 racks, 96 each
Catalog # 736-0759
Supplier: Thermo Fisher Scientific
Robotic tips for Eppendorf epMotion® automated pipetting system
Catalog # 736-0759
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
- Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
- Tips have a low percentage Coefficient of Variation (% CV)
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Documents
Specifications
- Confezione:10 racks, 96 each
- Sterili:+
- Volume:300 µl
- Filtro:-
- Cat. No.:736-0759
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)