Robotic tips for Eppendorf epMotion® automated pipetting system
50 µl, +, +, 10 racks, 96 each
Codice catalogo 736-0756
Nome del fornitore: Thermo Fisher Scientific
Robotic tips for Eppendorf epMotion® automated pipetting system
Codice catalogo 736-0756
Nome del fornitore: Thermo Fisher Scientific
Vai a
Product Details & Documents
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
- Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
- Tips have a low percentage Coefficient of Variation (% CV)
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Documents
Specifiche
- Confezione:10 racks, 96 each
- Sterili:+
- Volume:50 µl
- Filtro:+
- Cat. No.:736-0756
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..