Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Quick-RNA™ Kits

Quick-RNA™ Kits

Quick-RNA™ Microprep kit, 50 preps, 50
Codice catalogo ZYMOR1050
Marketsource
Nome del fornitore:   Zymo Research
Quick-RNA™ Kits
Codice catalogo ZYMOR1050
Nome del fornitore:  Zymo Research
Marketsource
Numero CAS:  
Quick-RNA™ Kits
Quick-RNA™ Kits

Quick-RNA™ Microprep kit, 50 preps, 50

Numero CAS:  
Price
SFr. 301.00
Log in per disponibilità
1 * 50 ST
Product Details & Documents

Isolation of RNA from cells and soft tissues.

  • DNA-free: Genomic DNA removal column and DNase I included
  • NGS-ready: RNA is ready for all downstream applications including next-gen sequencing, RT-qPCR, hybridisation and more

The quick-RNA kits are fast RNA isolation kits designed for the easy, reliable, and rapid isolation of DNA-free RNA from a wide range of cell and tissue samples. The procedure combines a unique buffer system with zymo-spin column technology to yield high quality total RNA (including small RNAs 17 to 200 nt) in about 10 minutes.

The procedure for these RNA isolation kits is simple and fast. Add the provided RNA lysis buffer to a sample, and then purify the RNA using the zymo-spin columns. The result is highly-concentrated, DNA-free RNA that is suitable for RT-PCR, hybridisation, sequencing and more.

In addition, these RNA isolation kits can be used for the enrichment of small and large RNAs into separate fractions.

Informazioni per l'ordine: This RNA isolation kit also includes DNase I.

Specifiche

Specifiche

  • Conf:
    50
  • Descrizione:
    Quick-RNA™ Microprep kit, 50 preps
  • Cat. No.:
    ZYMOR1050
  • Prep size:
    Micro
  • Refine target molecule:
    Total RNA
  • Target molecule:
    RNA
  • Method / Format:
    Column (silica-based)
  • Sample type:
    Cultured cells|Human/animal cells and tissues
  • Resa:
    Microprep: 10 µg RNA (binding capacity), ≥6 µl (elution volume)
  • Descrizione:
    Quick-RNA™ Microprep Kit
  • Capacità campioni:
    10 µg
  • Dimensione del campione:
    ≤1×10⁶ Cells or 5 mg Tissue

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.