Heating tapes
Heating tape, 3,1 m length, 230 W
Catalog # WINKWBS00203100230
Supplier: WINKLER
Jump to:
Product Details & Documents
These heating tapes contain four heat conductors, insulated silicone, outer braiding with tinned copper wire, factory terminated and vulcanised main connection.
- Very high and constant heating power per meter
- High operating temperature
- Extremely flexible
- Quick and easy installation
Specifications
- Descrizione:Heating tape, 3,1 m length, 230 W
- Seq:10
- Cat. No.:WINKWBS00203100230