2-Chlorophenylboronic acid
Glass bottle for solids, 100 g
Codice catalogo COBBBB-2259-100G
Marketsource
Nome del fornitore: COMBI-BLOCKS
2-Chlorophenylboronic acid
Codice catalogo COBBBB-2259-100G
Nome del fornitore: COMBI-BLOCKS
Marketsource
Numero CAS: 3900-89-8
Vai a
Product Details & Documents
Sinonimi: (2-Chlorophenyl)metaboric acid, Dihydroxy(2-chlorophenyl)borane, (2-Chlorophenyl)boronic acid, 2-Chlorobenzeneboronic acid
- Numero CAS:3900-89-8
- Formula:C₆H₆BClO₂
- Numero MDL:MFCD00674012
- Molecular Weight:156.38 g/mol
Specifiche
Specifiche
- Tipo di conf.:Glass bottle for solids
- Conf:100 g
Specifiche
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



