Film applicators, multi-functional
Multi-functional film applicator, 4 sided, gaps at 30, 60, 90, 120 µm, width 60 mm
Catalog # SIMEVF2169
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Multi-functional film applicator with 4 application sides for applying paint films of 4 different predefined thicknesses. The applicators are available in film widths of 60, 80 mm and one side of the applicator is supplied with a guidance support for straight application. Made from high grade stainless steel, the multi-functional film applicator will not be affected by acid or base elements.
Specifications
- Descrizione:Multi-functional film applicator, 4 sided, gaps at 30, 60, 90, 120 µm, width 60 mm
- Seq:03