Surface roughness comparators
Steel surface roughness comparator for shot blasting, Profiles: 25, 40, 70, 100
Katalognr. SIMELD2050
Anbieter: Industrial Physics
Direkt zu
Product Details & Documents
Quality steel comparison standards designed to indicate the surface condition of blasted steel, in grades of fine, medium and coarse.
- High purity electroformed nickel reference plates
- Copied from a certified mild steel master
- Protected test surfaces when not in use
Certifications: ASTM D4417, ISO 8503
Informations de livraison: Supplied in a sturdy leather pouch.
Documents
TQC Company Video
Vidéos
Technische Daten
- Modello:Profiles: 25, 40, 70, 100
- Descrizione:Steel surface roughness comparator for shot blasting