Surface profile comparators
Surface profile comparator for shot
Codice catalogo SIMELD2053
Nome del fornitore: Industrial Physics
Vai a
Product Details & Documents
Surface comparators for determining roughness by touch and sight.
- Five different grades of roughness in each comparator
- Designed to be used with the illuminated magnifier
- Available in Imperial units only
Section profiles: 0,5 ; 1,0 ; 2,0 ; 3,0 ; 4,0 mils for the three versions.
Certifications: ASTM D4417
Documents
TQC Company Video
Videos
Specifiche
- Descrizione:Surface profile comparator for shot
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Surface profile disc holder and magnifier (5×)Surface profile disc holder and magnifier (5×)SIMELD2054Log in per disponibilità1 * 1 ST - 151.00 SFr.