Surface roughness comparators
Surface roughness comparator for grit blasting, Profiles: 25, 60, 100, 150
Catalog # SIMELD2040
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Quality steel comparison standards designed to indicate the surface condition of blasted steel, in grades of fine, medium and coarse.
- High purity electroformed nickel reference plates
- Copied from a certified mild steel master
- Protected test surfaces when not in use
Certifications: ASTM D4417, ISO 8503
Delivery information: Supplied in a sturdy leather pouch.
Documents
TQC Company Video
Videos
Specifications
- Modello:Profiles: 25, 60, 100, 150
- Descrizione:Surface roughness comparator for grit blasting