Transfection reagent, Turbofect™
TurboFect™ transfection reagent, 200 µl, 200 µl
Codice catalogo PIERR0533
Nome del fornitore: Thermo Fisher Scientific
Transfection reagent, Turbofect™
Codice catalogo PIERR0533
Nome del fornitore: Thermo Fisher Scientific
Numero CAS:
Vai a
Product Details & Documents
TurboFect™ reagent an easy to use, non-immunogenic transfection reagent for delivery of DNA plasmids and expression vectors into eukaryotic cells.
- High transfection efficiency of a wide variety of cell types
- No optimisation required for most cell lines
- Excellent transfection efficiency in the presence or absence of serum
- Minimal cytotoxicity
- Ready to use without any need to reconstitute, dilute or manipulate
TurboFect™ reagent is ideal for stable and transient transfection or co-transfection of DNA or RNA, experiments with adherent or suspension cells, differentiated or undifferentiated cells and gene expression studies and assays involving GFP or luciferase reporters.
Specifiche
Specifiche
- Conf:200 µl
- Descrizione:TurboFect™ transfection reagent, 200 µl
- Cat. No.:PIERR0533
- DNA:Yes
Specifiche
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen..