LPT100 4-Leiter Temperaturfühler, -200...+150°C, DIN A (-70°..+150°C), dampffest, druckfest, autoclavierbar, D=5mm, LS=100mm, PTFE k abel L=1.2 m, ohne Stecker, T max. Kabel 220°C Einsatz im Sattdampf +150°C 1 * 1 Pack.
Codice catalogo ELPR800718
Nome del fornitore: ELPRO-BUCHS
Frequently Bought Together