Accessori
Parafiamma, All Rötzmeier safety canisters (5, 10 and 20 l) and safety barrels (10 and 25 l)
Catalog # 119-0169
Supplier: ROETZMEIER SICHERH BEHAELTER
Jump to:
Specifications
- Per:All Rötzmeier safety canisters (5, 10 and 20 l) and safety barrels (10 and 25 l)
- Descrizione:Parafiamma
- Seq:63