Serbatoi
Soaking tank for controlled moistening, soaking or bathing of particels, cloths, flannels, paper, etc., e.g. for cleaning with solvents, 4 l, Vasca di ammollo
Codice catalogo 119-0143
Nome del fornitore: ROETZMEIER SICHERH BEHAELTER
Vai a
Specifiche
- Tipo:Vasca di ammollo
- Capacità:4 l
- Descrizione:Soaking tank for controlled moistening, soaking or bathing of particels, cloths, flannels, paper, etc., e.g. for cleaning with solvents