Earplugs dispenser, Leight® Source 500
Earplugs refill Laser Lite, t-shape, SNR 35 dB, for dispenser Leight® Source 500, giallo/rosso
Codice catalogo DALL3301271
Nome del fornitore: HONEYWELL SAFETY
Earplugs dispenser, Leight® Source 500
Codice catalogo DALL3301271
Nome del fornitore: HONEYWELL SAFETY
Vai a
Product Details & Documents
The Leight Source 500 dispenser provides a highly visible permanent hearing protection source.
- Heavy-duty anodized aluminum allowing constant use
- Wall mounting for a user-friendly access
- Crank handle to dispense earplugs
Suitable for large factories and process industries with Max®, Max Lite®, Laser Lite®, Multi Max®, Matrix™, FirmFit™, Quiet® earplugs.
Specifiche
- Colore:giallo/rosso
- Descrizione:Earplugs refill Laser Lite, t-shape, SNR 35 dB, for dispenser Leight® Source 500
- Seq:02
- Cat. No.:DALL3301271
- Tipo di dispenser:Montabile a parete
- PPE type:Tappi auricolari