Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Safety helmets, Concept Roofer

Safety helmets, Concept Roofer

Safety helmet, unvented, with ratchet, EU, giallo
Codice catalogo 111-0870
Nome del fornitore:   CENTURION SAFETY PRODUCTS
Safety helmets, Concept Roofer
Codice catalogo 111-0870
Nome del fornitore:  CENTURION SAFETY PRODUCTS
Safety helmets, Concept Roofer
Safety helmets, Concept Roofer

Safety helmet, unvented, with ratchet, EU, giallo

Price
SFr. 30.20
Log in per disponibilità
1 * 1 ST
Product Details & Documents

The Concept Roofer is a light, strong and comfortable helmet made from ABS.

  • With premium ratchet headband ensuring secure fit
  • Six point terylene webbing cradle
  • Hydro-flock sweatband

Size: 520 to 630 mm

Weight: 340 g

EN 397

Certifications: EN 397

Documents

Specifiche
  • Colore:
    giallo
  • Descrizione:
    Safety helmet, unvented, with ratchet, EU
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.