Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Buffers, sodium phosphate buffer solutions (0,5 mol/L)

Buffers, sodium phosphate buffer solutions (0,5 mol/L)

Sodium phosphate stock solution (0,5 mol/L), 9,0, bottiglia in vetro, 500 ml
Catalog # CAYM600206-500
Supplier:   Cayman Chemical
Buffers, sodium phosphate buffer solutions (0,5 mol/L)
Catalog # CAYM600206-500
Supplier:  Cayman Chemical
CAS Number:  
Buffers, sodium phosphate buffer solutions (0,5 mol/L)

Sodium phosphate stock solution (0,5 mol/L), 9,0, bottiglia in vetro, 500 ml

CAS Number:  
Price
SFr. 166.00
Log in for availability
1 * 500 mL
Product Details & Documents
Specifications

Specifications

  • Tipo di conf.:
    bottiglia in vetro
  • Conf:
    500 ml
  • Descrizione:
    Sodium phosphate stock solution (0,5 mol/L)
  • Valore pH:
    9,0

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.