Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Buffers, sodium phosphate buffer solutions (0,5 mol/L)

Buffers, sodium phosphate buffer solutions (0,5 mol/L)

Sodium phosphate stock solution (0,5 mol/L), 8,0, bottiglia in vetro, 250 ml
Codice catalogo CAYM600205-250
Nome del fornitore:   Cayman Chemical
Buffers, sodium phosphate buffer solutions (0,5 mol/L)
Codice catalogo CAYM600205-250
Nome del fornitore:  Cayman Chemical
Numero CAS:  
Buffers, sodium phosphate buffer solutions (0,5 mol/L)

Sodium phosphate stock solution (0,5 mol/L), 8,0, bottiglia in vetro, 250 ml

Numero CAS:  
Price
87.60 SFr.
Log in per disponibilità
1 * 250 mL
Product Details & Documents
Specifiche

Specifiche

  • Tipo di conf.:
    bottiglia in vetro
  • Conf:
    250 ml
  • Descrizione:
    Sodium phosphate stock solution (0,5 mol/L)
  • Valore pH:
    8,0

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.