Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
2-Methoxyestradiol

2-Methoxyestradiol

Bottiglia di vetro, 50 mg
Catalog # CAYM13021-50
Supplier:   Cayman Chemical
2-Methoxyestradiol
Catalog # CAYM13021-50
Supplier:  Cayman Chemical
CAS Number:  362-07-2
2-Methoxyestradiol

Bottiglia di vetro, 50 mg

CAS Number:  362-07-2
Price
SFr. 452.00
Log in for availability
1 * 50 mg
Product Details & Documents
Synonyms:  (8R,9S,13S,14S,17S)-2-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
  • CAS Number:362-07-2
  • Formula:C₁₉H₂₆O₃
  • MDL Number:MFCD00010489
  • Molecular Weight:302.408 g/mol
Specifications

Specifications

  • Tipo di conf.:
    Bottiglia di vetro
  • Conf:
    50 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.