Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Tetranor PGDM ≥90%

Tetranor PGDM ≥90%

100 µG
Codice catalogo CAYM12850-100
Nome del fornitore:   Cayman Chemical
Tetranor PGDM ≥90%
Codice catalogo CAYM12850-100
Nome del fornitore:  Cayman Chemical
Numero CAS:  70803-91-7
Tetranor PGDM ≥90%

100 µG

Numero CAS:  70803-91-7
Price
SFr. 1’120.00
Log in per disponibilità
1 * 100 µG
Product Details & Documents

Tetranor PGDM is a major metabolite of prostaglandin D2 which are mainly involved in the inhibition of platelet aggregation and relaxation of vascular smooth muscle.

  • Formulated as a solution in methanol

The levels of tetranor PGDM is found to be as 1,5±0,3 ng/mg creatinine and 8,1±1,3 ng/mg creatinine in human and mouse urine respectively.

Sinonimi:  tetranor-Prosatglandin D Metabolite, tetranor-PGD Metabolite
  • Numero CAS:70803-91-7
  • Formula:C₁₆H₂₄O₇
  • Molecular Weight:328.4 g/mol
  • temperatura di conservazione:Ultracongelamento (–80°C)

Documents

Specifiche

Specifiche

  • Conf:
    100 µG

Specifiche

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.