Chenodeoxycholic acid
Glass bottle for solids, 25 g
Catalog # CAYM10011286-25
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 3α,7α-Dihydroxycholanic acid, 7α-Hydroxylithocholic acid, 7alpha-Hydroxylithocholic acid, 3α,7α-Dihydroxy-5β-cholan-24-oic acid, 3α,7α-Dihydroxycholan-24-oic acid, Chenodiol, 3α,7α-Dihydroxy-5β-cholanic acid
- CAS Number:474-25-9
- Formula:C₂₄H₄₀O₄
- MDL Number:MFCD00064142
- Molecular Weight:392.58 g/mol
- Merck Index:12,02096
- Melting Pt:164,85…165.85 °C
- Density:1,162 g/cm³ (20 °C)
- :Density: 1.162 g/cm³ (20 °C)
- EINECS:207-481-8
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:25 g
- Cat. No.:CAYM10011286-25
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



