(R)-(-)-Mandelic acid
Glass bottle for solids, 100 g
Catalog # CAYM10007128-100
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: D(-)-Mandelic acid, (R)-(-)-2-Hydroxy-2-phenylacetic acid, (R)-2-Hydroxy-2-phenylacetic acid, D(-)-2-Hydroxy-2-phenylacetic acid
- CAS Number:611-71-2
- Formula:C₈H₈O₃
- MDL Number:MFCD00064251
- Molecular Weight:152.15 g/mol
- Boiling Pt:322 °C (1013 hPa)
- Melting Pt:130…132 °C
- Density:1,341 g/cm³ (20 °C)
- Flash Pt:>190 °C
- EINECS:210-276-6
Specifications
Specifications
- Tipo di conf.:Glass bottle for solids
- Conf:100 g