Supports for stand grids
Flat flanges LF 150, framework grid 0,500 m
Codice catalogo LENZ06247416
Nome del fornitore: Lenz Laborglas
Vai a
Product Details & Documents
Stainless steel (clamp), steel, powder coated (supports).
For fixing lids onto reaction vessels and for the suspension into stands, complete with clamp and supports.
Specifiche
- Per:Flat flanges LF 150, framework grid 0,500 m
- Seq:02