Multimodal chromatography media, Capto™ Core 700
Capto™ Core 700, Liquid, 90 µm, 25 ml
Codice catalogo 17-5481-01
Nome del fornitore: Cytiva
Multimodal chromatography media, Capto™ Core 700
Codice catalogo 17-5481-01
Nome del fornitore: Cytiva
Numero CAS:
Vai a
Product Details & Documents
Capto™ Core 700 is designed for intermediate purification and polishing of viruses and other large biomolecules (molecular weight >700000).
- Novel core bead technology allows efficient capture of contaminants while target molecules are collected in the flowthrough
- Significantly improved productivity and higher flow rates compared with gel filtration methods
- Straightforward optimisation due to flowthrough chromatography and robust performance
- Convenient small-scale purification, process development, and scale-up using prepacked HiTrap and HiScreen columns
The novel core bead technology and multimodal, octylamine ligand give dual functionality, size exclusion, and binding chromatography in one chromatography medium (resin). Available in various pack sizes for packing columns and in convenient, prepacked formats.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Ulteriori dettagli sul prodotto
Specifiche
Specifiche
- Dimensioni particelle:90 µm
- Conf:25 ml
- Descrizione:Capto™ Core 700
- Tipo:Liquid
- Cat. No.:17-5481-01
- Capacità legante:~ 13 mg ovalbumin
- Matrice:Highly cross-linked agarose
- Portata:<500 cm/h
- Range di pH:3 - 13
- Capacità ionica totale:40 - 85 µmol/ml (Cl-)
- Ligando:Octylamine
- Temperatura di conservazione:4 - 30 °C



