Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Zymo-Spin™ IC Columns

Zymo-Spin™ IC Columns

Zymo-Spin™ IC, Spin Column, 250 pack, 250
Catalog # ZYMOC1004-250
Marketsource
Supplier:   Zymo Research
Zymo-Spin™ IC Columns
Catalog # ZYMOC1004-250
Supplier:  Zymo Research
Marketsource
CAS Number:  
Zymo-Spin™ IC Columns
Zymo-Spin™ IC Columns

Zymo-Spin™ IC, Spin Column, 250 pack, 250

CAS Number:  
Price
SFr. 328.00
Log in for availability
1 * 250 ST
Product Details & Documents

Spin columns for the purification of DNA or RNA.

  • Format: DNA/RNA binding
  • DNA binding capacity: 5 µg
  • RNA binding capacity: 10 µg
  • Compatibility: microcentrifuge, vacuum manifold

Capped version of the Zymo-Spin™ I column. The Zymo-Spin™ IC column can be used either in microcentrifuges or on vacuum manifolds for the purification of DNA and/or RNA. The Zymo-Spin™ IC features durable polypropylene construction and contains a unique silica-based matrix that allows purification of up to 5 μg of DNA, and 10 μg of RNA, in ≥ 6 μl eluate. Capacity is 800 μl.

Specifications

Specifications

  • Tipo di conf.:
    250 pack
  • Conf:
    250
  • Descrizione:
    Zymo-Spin™ IC
  • Formato:
    Spin Column

Specifications