Orders LinkContactUsLinkComponent
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography anion multi-element standard solution V, (6 elements in water) 10 mg/l: F, Br / 50 mg/l: NO₃, PO₄ / 100 mg/l: Cl / 200 mg/l: SO₄, Certipur®, 100 mL
Codice catalogo 1.09032.0100
Nome del fornitore:   Merck
Ion chromatography Multi-Element standards, Supelco®
Codice catalogo 1.09032.0100
Nome del fornitore:  Merck
Numero CAS:  
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography anion multi-element standard solution V, (6 elements in water) 10 mg/l: F, Br / 50 mg/l: NO₃, PO₄ / 100 mg/l: Cl / 200 mg/l: SO₄, Certipur®, 100 mL
Numero CAS:  
Price
SFr. 444.00
Log in per disponibilità
1 * 100 mL
Product Details & Documents
All ion chromatography standards are traceable to NIST standard reference materials.
Specifiche
Specifiche
  • Conf:
    100 mL
  • Descrizione:
    Ion chromatography anion multi-element standard solution V, (6 elements in water) 10 mg/l: F, Br / 50 mg/l: NO₃, PO₄ / 100 mg/l: Cl / 200 mg/l: SO₄, Certipur®