Orders LinkContactUsLinkComponent
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography cation multi-element standard solution I, (5 elements in diluted nitic acid) 10 mg/l: NH₄ / 50 mg/l: K / 100 mg/l: Na, Ca, Mg, Certipur®, 100 mL
Codice catalogo 1.09036.0100
Nome del fornitore:   Merck
Ion chromatography Multi-Element standards, Supelco®
Codice catalogo 1.09036.0100
Nome del fornitore:  Merck
Numero CAS:  
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography Multi-Element standards, Supelco®
Ion chromatography cation multi-element standard solution I, (5 elements in diluted nitic acid) 10 mg/l: NH₄ / 50 mg/l: K / 100 mg/l: Na, Ca, Mg, Certipur®, 100 mL
Numero CAS:  
Price
SFr. 287.00
Log in per disponibilità
1 * 100 mL
Product Details & Documents
All ion chromatography standards are traceable to NIST standard reference materials.
Specifiche
Specifiche
  • Conf:
    100 mL
  • Descrizione:
    Ion chromatography cation multi-element standard solution I, (5 elements in diluted nitic acid) 10 mg/l: NH₄ / 50 mg/l: K / 100 mg/l: Na, Ca, Mg, <sub> </sub>Certipur®