Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Provette per centrifuga

Provette per centrifuga

Tubes, with blue screw cap, 15 ml, 16×100 mm, Tappo a vite, blu, non sterile
Catalog # KART8830304
Supplier:   KARTELL
Provette per centrifuga
Catalog # KART8830304
Supplier:  KARTELL
Provette per centrifuga
Provette per centrifuga

Tubes, with blue screw cap, 15 ml, 16×100 mm, Tappo a vite, blu, non sterile

Price
SFr. 298.00
Log in for availability
1 * 1.000 ST
Product Details & Documents

PP with quick closure PE screw cap.

  • Leak-proof
  • Can be centrifuged up to 4000×g
  • Cap is designed to avoid any contamination due to drops of liquid on the inner surface
Specifications
  • Tipo di chiusura:
    Tappo a vite
  • Ø×L:
    16×100 mm
  • Versione:
    non sterile
  • Capacità:
    15 ml
  • Descrizione:
    Tubes, with blue screw cap
  • Seq:
    03
  • Colore tappo:
    blu
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.