Orders LinkContactUsLinkComponent
Closures for headspace vials
Closures for headspace vials
White Ultra bleed PTFE coated silicone skived pressure relief aluminium cap, pre-assembled
Catalog # PERCN9302975
Supplier:   PerkinElmer
Closures for headspace vials
Catalog # PERCN9302975
Supplier:  PerkinElmer
Closures for headspace vials
White Ultra bleed PTFE coated silicone skived pressure relief aluminium cap, pre-assembled
Price
1 890.00 SFr.
Log in for availability
1 * 1.000 ST
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Specifications
  • Descrizione:
    White Ultra bleed PTFE coated silicone skived pressure relief aluminium cap, pre-assembled
  • Seq:
    19