Double spatulas, powder
210 mm, 9 mm
Codice catalogo 231-0546
Nome del fornitore: RSG
Vai a
Product Details & Documents
Stainless steel.
- With round handle, one flat and one scoop shaped, rounded spatula end
- Straight shape
- Very good thermal resistance, autoclavable
- Good chemical resistance
Specifiche
- Lunghezza:210 mm
- Larghezza lama:9 mm