Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
ChIP Kit - Plants, Abcam

ChIP Kit - Plants, Abcam

ChIP Kit - Plants
Catalog # ABCAAB117137-24
Marketsource
Supplier:   Abcam
ChIP Kit - Plants, Abcam
Catalog # ABCAAB117137-24
Supplier:  Abcam
Marketsource
CAS Number:  
ChIP Kit - Plants, Abcam

ChIP Kit - Plants

CAS Number:  
Price
SFr. 529.00
Log in for availability
1 * 24 Tests
Product Details & Documents

Protein-DNA interactions play a critical role for cellular functions such as signal transduction, gene transcription and epigenetic silencing.

Caution: All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Specifications

Specifications

  • Descrizione:
    ChIP Kit - Plants
  • Cat. No.:
    ABCAAB117137-24
  • Temperatura di conservazione:
    Store at Multi.

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.