Mop heads, DryMop™
Dry Mop™ frame, heavy-gauge metal with zinc-plating, 1219 mm
Codice catalogo 129-0606
Nome del fornitore: MICRONOVA
Vai a
Product Details & Documents
For dusting and dry mopping large surface areas.
- Low profile design
- Polyester pad head limits particle generation
- Swivel head for easy maneuvering
- Launderable/Autoclavable covers
Specifiche
- Descrizione:Dry Mop™ frame, heavy-gauge metal with zinc-plating
- Lunghezza:1219 mm
- Seq:03