Pycnometers for bituminous materials
Bottle with numbering and matt shield, without top, borosilicate glass 3.3, 1000 ml, 60/46
Codice catalogo NEUB0321-50-0011
Nome del fornitore: NEUBERT VOLUME GLASSWAERE
Pycnometers for bituminous materials
Codice catalogo NEUB0321-50-0011
Nome del fornitore: NEUBERT VOLUME GLASSWAERE
Vai a
Product Details & Documents
Soda-lime glass or borosilicate glass 3.3.
- Two-part assembly both parts with identification number
- Cap with funnel for ease of filling
- Ring mark on cap stem
Specifiche
- Cono (NS):60/46
- Volume (nom.):1000 ml
- Descrizione:Bottle with numbering and matt shield, without top, borosilicate glass 3.3
- Seq:05