Test tubes with screw caps
10 ml, 100 mm, 16 mm, In bulk, Non sterili
Codice catalogo KART88200
Nome del fornitore: KARTELL
Vai a
Product Details & Documents
PS, clear, wtih screw cap.
- Tight screw cap
- Good chemical resistance
- Good temperature resistance
Taking into consideration the different applications, they are available sterile single packed, sterile nultiple packed and not sterile.
Specifiche
- Ø est:16 mm
- Confezione:In bulk
- Sterilità:Non sterili
- Capacità:10 ml
- Lunghezza:100 mm