Multi-parameter meter (pH/ISE/conductivity/oxygen), bench top, Orion™ Versa Star Pro™
Product Details & Documents
These bench top meters with pH/ISE, conductivity and Dissolved Oxygen/RDO modules meet the most challenging applications for pH, ion concentration, conductivity, dissolved oxygen and temperature. The meters offer interchangeable measurement modules that allow multiple users to customise four separate channels to meet their specific requirements.
- Colour display with adjustable text sizes; numeric keypad; onscreen measurement stability indicator
- Auto-Read to hold a stable reading; ten password-protected methods per channel
- Audible alarms automatically notify users when a set offset value, high/low limit value, and calibration time interval are reached
- Uniformly mix solutions using up to two meter-controlled stirrer probes that are easy to position and can be quickly rinsed between samples
- Data log collects and date/time stamps up to 2000 measurement sets with optional electrode ID, sample ID and account ID
- Calibration log stores ten most recent calibrations per parameter with viewable graph for multipoint pH and ISE calibrations
- Meter facilitates data transfers to a computer via USB or RS232 connections
IP 54-rated housing resists dust and splashes and can be wall-mounted if table space is limited.
When used in conjunction with the Orion Navigator Pro software, the Orion Versa Star Pro meter is designed to create electronic data records and audit trails with password-protection, which can be used to assist with 21 CFR Part 11 requirements when paired with each laboratory’s proper IT protocols, SOPs and training records.*Orion Navigator Pro software offers multi-level system access for an administrator and up to 10 users. The system administrator can customize each user’s access level from full system access to basic data collection only. The Navigator Pro software controls all meter functions including meter settings, calibration, measurement, and reporting to help ensure data integrity and provide a complete audit trail.
Features and benefits
- Generates electronic records and signatures
- Creates audit trail report for electronic records
- Limits access to authorized individuals
- Password protection with system administrator and up to 10 users
- Full computer control of meter functions including setup, calibration and measurement
- Paperless record keeping system
: All meters are supplied with one or two VSTAR-ISE pH/ISE modules, one VSTAR-CND conductivity module, one VSTAR-RD RDO/DO module, electrode stand, universal power adapter, USB cable, literature CD, printed quick start guide, computer interface cable and meter test certificate.
Kits additionally include: 8157BNUMD ROSS Ultra Triode epoxy-body pH/ATC electrode, 013005MD Orion DuraProbe 4-cell (K=0,475) conductivity sensor, 083005MD Orion polarographic dissolved oxygen sensor, 096019 Orion Star stirrer probe, 810199 ROSS pH buffer and solution kit, 011007 Orion 1413 µS conductivity standard, 080513 DO maintenance kit, BOD adapter/funnel/stirrer.
: * It is the responsibility of each laboratory to validate the Navigator Pro computer software for its intended use, including all regulatory requirements.
Specifiche
- Descrizione:Multi-parameter meter, bench top, Orion™ Versa Star™ Pro™ kit, with 1 × pH/ISE, 1 × conductivity and 1 × RDO/DO module
- Seq:03
- Precisione della conducibilità:0,5% of reading ±1 digit (> 3 μS); 0,5% of reading ±0,01 μS (≤ 3 μS)
- Salinità - accuratezza:0,5% of reading ±1 digit
- pH - risoluzione:0,1/0,01/0,001
- ISE unità:ppm, M, mg/L, %, ppb o senza unità
- Salinità - range:0,06 - 80,00 psu/0,05 - 42,00 ppt
- Capacità della memoria:2000 punti
- Lingue:English, Spanish, French, Italian, German, Chinese, Portuguese and Korean
- Range di temperatura:−5,0...+105,0 °C
- ISE calibrazione:fino a 6
- Peso:1,2 kg (modules add 0,2 kg each)
- Modello:Orion™ Versa Star Pro™
- Datalog - funzione di registrazione:Automatic data logging with Auto-Read, Timed and Single Shot measure modes; manual data logging with Continuous measure mode
- OD concentrazione:DO: 0,00 - 90,00 mg/L; RDO: 0,00 - 50,00 mg/L
- Uscite:RS-232, USB
- mV - range:±2000
- Salinità - risoluzione:0,01 psu or 0,01 ppt minimum, auto ranging
- TDS - accuratezza:0,5% of reading ±1 digit
- Calibrazione della temperatura:1 point offset
- Calibrazione della conducibilità:fino a 6
- Interfacce:BNC (pH or ISE), 8-pin Mini-DIN (temperature), reference pin-tip, 8-pin Mini-DIN (conductivity with temperature probe), 9-pin Mini-DIN (RDO or polarographic DO with temperature probe), 2 stirrer inputs
- ISE - gamma:0,0001 - 19990 ppm
- Risoluzione della temperatura:0,1 °C
- Intervallo di conducibilità:0,00 - 3000 mS/cm
- pH - calibrazione:fino a 6 punti
- TDS - fattore di conversione:Linear (0,02 - 9,99)
- Resistività - accuratezza:0,5% of reading ±1 digit
- Cat. No.:ORIOVSTAR92
- Range della temperatura di esercizio:5...45 °C (ambient)
- mV - accuratezza:±0,2 mV o ±0,05% del valore, in base al maggiore
- Alimentazione:Universale, adattatore di alimentazione c.a., 90 - 260 V c.a., 50 - 60 Hz
- Calibrazione:pH with calibration editing option, ion concentration (ISE) with calibration editing option, relativ
- Accuratezza della temperatura:±0,1 °C
- L×P×A:280×240×100 mm
- mV - risoluzione:0,1
- Temperatura - compensazione:Linear, nLFn (non-linear ultra pure water), nLFu (non-linear ultra pure degassed water), EP (off with warning) or off
- pH - range:Da -2 a +20
- OD - range di pressione barometrica:400 - 850 mmHg
- TDS - risoluzione:0,001 - 200,0 ppm
- Costante di cella di conducibilità:0,001 - 199,9 cm⁻¹
- Punti di calibrazione:1 point relative mV (RmV) mode, 1 point ORP mode
- OD accuratezza:DO: ±0,2 mg/l or ±2% of reading (whichever is greater)
RDO: ±0,1 mg/l (up to 8 mg/l), ±0,2 mg/l (8 - 20 mg/l), ±10% of reading (up to 50 mg/l)
±2 % saturation or ±2% of reading whichever is greater - ISE accuratezza:±0,2 mV o ±0,5%, in base al maggiore
- mV - calibrazione:1 point
- OD - saturazione:0,0 - 600,0 %
- Conformità:IP 54
- Risoluzione della conducibilità:0,01 µS minimo; minimo 4 cifre significative
- Risoluzione della resistività:1 Ω/cm/0,1 MΩ/cm
- ISE risoluzione:0,0001 minimum, 1 to 4 significant digits (user selectable)
- OD - risoluzione:0,01/0,1 mg/L
- N. di canali:4 - customisable with modules
- pH - accuratezza:±0,002
- Display:Colour graphic LCD with backlight
- Resistività - range:2 Ω/cm - 100,0 MΩ/cm
Services
Informazioni sul prodotto
- Codice catalogoDescrizioneDisponibilitàPrice
- ORIOIQOQ-VSTARIQ/OQ documentation for Orion™ Versa Star metersLog in per disponibilità1 * 1 ST - SFr. 433.00
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Universal power adapter for Orion™ Versa Star™ meters, with US, EU, UK and China plug, 90 - 260 VAC, 50/60 HzUniversal power adapter for Orion™ Versa Star™ meters, with US, EU, UK and China plug, 90 - 260 VAC, 50/60 HzORIOSTARA-PWRLog in per disponibilità1 * 1 ST - SFr. 81.10



